Learn More
Aviva Systems Biology Corp NEUROD1 Antibody - middle region (ARP38162_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38162_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEUROD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NEUROD1. Alternate names: BETA2, BHF-1, NEUROD, MODY6, bHLHa3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Sheep, Zebrafish
Catalog No. 50-014-56546