Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology NeuroD1 Monoclonal Antibody, Rabbit IgG, Unconjugated, 100 μL
SDP

Catalog No. 502732453
Encompass

This NeuroD1 (E3E4F) rabbit monoclonal antibody recognizes endogenous levels of total NeuroD1 protein. NeuroD1 is a member of the basic helix-loop-helix (bHLH) family of transcription factors that plays a crucial role in development, including the pancreas, nervous system, inner ear, and retina.

  • Recombinant rabbit monoclonal antibody
  • Validated for Western blotting, immunoprecipitation, and immunofluorescence applications
  • Species reactivity: human, mouse, and rat
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide
  • Store at −20°C

Catalog No. 50-273-2453 Supplier Cell Signaling Technology Supplier No. 62953S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This NeuroD1 (E3E4F) rabbit monoclonal antibody recognizes endogenous levels of total NeuroD1 protein. NeuroD1 is a member of the basic helix-loop-helix (bHLH) family of transcription factors that plays a crucial role in development, including the pancreas, nervous system, inner ear, and retina.

  • Recombinant rabbit monoclonal antibody
  • Validated for Western blotting, immunoprecipitation, and immunofluorescence applications
  • Species reactivity: human, mouse, and rat
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide
  • Store at −20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.