Learn More
Aviva Systems Biology Corp NEUROD4 Antibody - middle region (ARP32415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEUROD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NEUROD4. Alternate names: ATH-3, Atoh3, MATH-3, ATH3, bHLHa4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep, Zebrafish
Catalog No. 50-014-54579