Learn More
Cayman Chemical Neurodazine 10mg
SDP
Catalog No. 502951334
Encompass_Preferred
Induces neuronal differentiation in skeletal muscle cells, as indicated by the upregulated expression of neuron-specific markers; effective with mature muscle fibers as well as myoblasts
Catalog No. 50-295-1334 Supplier Cayman Chemical Supplier No. 1322410 MG
Induces neuronal differentiation in skeletal muscle cells, as indicated by the upregulated expression of neuron-specific markers; effective with mature muscle fibers as well as myoblasts