Learn More
Medchemexpress LLC HY-108439 10mg Medchemexpress, Neurodazine CAS:937807-66-4 Purity:>98%
SDP
Catalog No. 501968548
Encompass_Preferred
Medchemexpress, HY-108439 10mg Neurodazine CAS:937807-66-4 Neurodazine is an imidazole-based small molecule, serve as a promoter of neurogenesisin pluripotent cells. Neurodazine promotes neurogenesis by activating Wnt and Shh signaling pathways. Neurodazine selectively suppresses astrocyte differentiation of P19 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8548 Supplier Medchemexpress LLC Supplier No. HY10843910MG
Medchemexpress, HY-108439 10mg Neurodazine CAS:937807-66-4 Neurodazine is an imidazole-based small molecule, serve as a promoter of neurogenesisin pluripotent cells. Neurodazine promotes neurogenesis by activating Wnt and Shh signaling pathways. Neurodazine selectively suppresses astrocyte differentiation of P19 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.