Learn More
Abcam Neurodazine, Induces neurogenesis in skeletal muscle cells, 10MG
Catalog No. 502707161
Encompass_Preferred
MW 456.93 g/mol, Purity >98%. Induces neurogenesis in skeletal muscle cells including immature, differentiated myotubes (C2C12) and mature skeletal muscle. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7161 Supplier Abcam Supplier No. AB12089310MG
MW 456.93 g/mol, Purity >98%. Induces neurogenesis in skeletal muscle cells including immature, differentiated myotubes (C2C12) and mature skeletal muscle. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement