Learn More
Enzo Life Sciences Neurofilament H (non-phosphorylated) monoclonal antibody (SMI32) (purified) (100 µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2190100
Encompass_Preferred
Clone: SMI-32. Host: Mouse. Isotype: IgG1 UniProt: P12036 (human). Specificity: Recognizes a non-phosphorylated epitope in neurofilament H of most mammalian species. Species Reactivity: Mammalian. Applications: ELISA, ICC, IHC, WB. Purity: Purified. Formulation: Liquid. In PBS.
Catalog No. NC1166494