Learn More
Biogenex Laboratories Neurofilament : NE-14 : 6mL
SDP
Supplier:  Biogenex Laboratories AM0735M
Encompass_Preferred
Neurofilaments (10 nm diameter) and microtubules (25 nm diameter)comprise the main structural elements of neuronal axons, dendrites, andperikerya. Neurofilaments are composed of three major polypeptidesreferred to as the neurofilament triplet with approximate molecularweights of 200 kD, 160 kD and 68 kD. This antibody can be used forpositive identification of neurons in the central and peripheral nervoussystems. In general, co-expression of keratin and neurofilament should beinterpreted as indicating neuroendocrine differentiation of a given tissue orneoplasm
Catalog No. 50-197-8816