Learn More
Aviva Systems Biology Corp Neurog2 Antibody - N-terminal region (ARP37020_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Neurog2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Atoh4, Math4A, ngn-2, ngn2, bHLHa8ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-284-566