Learn More
Aviva Systems Biology Corp NEUROG2 Antibody - N-terminal region (ARP39585_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39585_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEUROG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NEUROG2. Alternate names: Atoh4, MGC46562, Math4A, NGN2, ngn-2, bHLHa8ApplicationWB. Species reactivity: Cow, Dog, Human, Rabbit
Catalog No. 50-014-57095