Learn More
Aviva Systems Biology Corp Neurog2 Antibody - C-terminal region (ARP39586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Neurog2. It was validated on Western Blot. . Alternate names: Atoh4, Math4A, bHLHa8, ngn-2, ngn2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-817