Learn More
Biomatik Corporation Neurogranin (Nrgn) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30401
Encompass_Preferred
Neurogranin (Nrgn) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Neurogranin (NRGN). The Neurogranin (Nrgn) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Met1~Ala67. Applications: WB, IHC. Target Synonyms: Neurogranin (NRGN). Accession Number: Q92686. Gene ID: 4900. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9206