Learn More
Enzo Life Sciences [beta-Ala8] Neurokinin A (4-10) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP5060001
Encompass_Preferred
Highly selective NK2 neurokinin receptor agonist. Purity: ≥97%. Solubility: Dilute acetic acid in H2O. Long Term Storage: -20°C.
Catalog No. 50-201-1357