Learn More
Medchemexpress LLC HY-P0236A 1mg Medchemexpress, Neurokinin A(4-10) (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0236A1MG
Encompass_Preferred
Medchemexpress, HY-P0236A 1mg Neurokinin A(4-10) (TFA) CAS: Neurokinin A (4-10) TFA is a tachykinin NK 2 receptor agonist [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4067