Learn More
Medchemexpress LLC HY-P0197A 5mg Medchemexpress, Neurokinin A (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0197A5MG
Encompass_Preferred
Medchemexpress, HY-P0197A 5mg Neurokinin A (TFA) CAS: Neurokinin A TFA (Substance K TFA), a peptide neurotransmitter of the tachykinin family, acts via the NK-2 receptor. Neurokinin A acts as a major mediator in human airway and gastrointestinal tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3027