Learn More
CPC Scientific H-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP033A
Encompass
Bombesin-like peptide isolated from porcine spinal cord that exhibits a potent stimulant effect on the smooth muscle of rat uterus.ONE-LETTER SEQUENCE: GNHWAVGHLM-NH2MOLECULAR FORMULA: C50H73N17O11S1MOLECULAR WEIGHT:1120.30STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [81608-30-2], SYNONYMS: GRP (18-27) (human, porcine, canine)RESEARCH AREA: NeuropeptidesREFERENCES: K.A. Roth et al., BBRC, 112, 528 (1983); N. Minamino et al., BBRC, 119, 14 (1984)
Catalog No. 50-210-9917