Learn More
CPC Scientific H-Phe-Lys-Val-Asp-Glu-Glu-Phe-Gln-Gly-Pro-Ile-Val-Ser-Gln-Asn-Arg-Arg-Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific NEUP035A
Encompass
Potent stimulator of rat uterus smooth muscle that has also been reported to influence blood pressure in rats and dogs.ONE-LETTER SEQUENCE: FKVDEEFQGPIVSQNRRYFLFRPRN-NH2MOLECULAR FORMULA: C144H217N43O37MOLECULAR WEIGHT:3142.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [98395-76-7], SYNONYMS: Neuromedin U-25 (porcine)RESEARCH AREA: NeuropeptidesREFERENCES: N. Minamino et al., BBRC, 130, 1078 (1985)
Catalog No. 50-210-9918