Learn More
Bachem Neuromedin U-23 (rat) Trifluoroacetate
SDP
Supplier:  Bachem 4016663.0001
Encompass_Preferred
Neuromedin U-23 (rat) Trifluoroacetate, H-Tyr-Lys-Val-Asn-Glu-Tyr-Gln-Gly-Pro-Val-Ala-Pro-Ser-Gly-Gly-Phe-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2, 117505-80-3, C124H180N34O31, Mr 2643.00, 1mg. NMU-23. rNMU-23 was isolated from the small intestine and exerts two-fold potent uterus stimulant activity compared to pig neuromedin U-25.
Catalog No. 50-260-236