Learn More
Carbosynth LLC. Neuromedin U (Mouse) (0.5 mg vial), 1ea, MW: 2706, Molecular formula: C125H185N35O33
SDP
Supplier:  Carbosynth LLC. PNM4491V0.5MG
Encompass_Preferred
Synonyms: NMU-23 (Mouse) Phe-Lys-Ala-Glu-Tyr-Gln-Ser-Pro-Ser-Val-Gly-Gln-Ser-Lys-Gly-Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2, Purity: , ≥99.0% (HPLC), Application: Food Intake Suppressor, Storage Temperature: -20 °C, References: P.J. Brighton, P.G. Szekeres, and G.B. Willars, Pharmacol. Rev., 56, 231 (2004). (Review), V.G. Martinez and L. O'Driscoll, Clin. Chem., 61, 471 (2015). (Review)
Catalog No. 50-168-6694