Learn More
CPC Scientific H-Tyr-Lys-Val-Asn-Glu-Tyr-Gln-Gly-Pro-Val-Ala-Pro-Ser-Gly-Gly-Phe-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP036B
Encompass
Peptide isolated from the small intestine that exerts uterus stimulant activity two-folds as potent when compared to pig neuromedin U25.ONE-LETTER SEQUENCE: YKVNEYQGPVAPSGGFFLFRPRN-NH2MOLECULAR FORMULA: C124H180N34O31MOLECULAR WEIGHT:2643.00STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117505-80-3], SYNONYMS: Neuromedin U-23 (rat), NMU-23RESEARCH AREA: NeuropeptidesREFERENCES: N. Minamino et al., BBRC, 156, 685 (1984)
Catalog No. 50-210-9921