Learn More
FUJIFILM Biosciences Neuron Dissociation Solutions
Catalog No. 5000450395
Encompass_Preferred
This is dispersion medium for primary neuronal cell cultures of rat and mouse and designed for the gentle and efficient isolation of neurons from neural tissue supporting downstream cell culture applications Please use this medium for dispersion of frozen nerve cells or fresh brain tissues dissected from rat and mouse This contains 4 sets of 3 different solutions of 5ml each enzyme solution dispersion solution and isolation solution
Catalog No. 50-004-50395 Supplier FUJIFILM Biosciences Supplier No. 29178001
This is dispersion medium for primary neuronal cell cultures of rat and mouse and designed for the gentle and efficient isolation of neurons from neural tissue supporting downstream cell culture applications Please use this medium for dispersion of frozen nerve cells or fresh brain tissues dissected from rat and mouse This contains 4 sets of 3 different solutions of 5ml each enzyme solution dispersion solution and isolation solution