Learn More
Neuromics, Inc Neuron Growth Media, 500 ml is a complete media that is composed of basal medium supplemented with 10% FBS and growth factors.
Catalog No. NC2071965
Encompass
Human Neuron Cells Growth medium (HNM 001) is a complete media that is composed of HNCs Basal Medium that has been supplemented with 10% high quality Fetal Bovine Serum and growth factors. It contains the essential nutrients for HNCs proliferation.
Catalog No. NC2071965 Supplier Neuromics, Inc Supplier No. HNM001
Human Neuron Cells Growth medium (HNM 001) is a complete media that is composed of HNCs Basal Medium that has been supplemented with 10% high quality Fetal Bovine Serum and growth factors. It contains the essential nutrients for HNCs proliferation.