Learn More
Diagnostic Biosystems NEURON SPECIFIC ENOLASE Neuron Specific Enolase
Catalog No. 502407613
Encompass_Preferred
This antibody reacts with human isoenzymes of neuron specific enolase containing γ subunits. This antibody gives a specific staining in neurons of the brain, in islets of Langerhansin pancreas and in peripheral nerves of the skin. No staining with other tissue types has been seen.
Catalog No. 50-240-7613 Supplier Diagnostic Biosystems Supplier No. PDR006
This antibody reacts with human isoenzymes of neuron specific enolase containing γ subunits. This antibody gives a specific staining in neurons of the brain, in islets of Langerhansin pancreas and in peripheral nerves of the skin. No staining with other tissue types has been seen.