Learn More
Sciencell Research Laboratories NEURONAL MEDIUM - 500 ML
Catalog No. NC0268695
Encompass
Neuronal medium is a complete medium designed for optimal growth of normal human neurons in vitro. It is shipped as a kit with neuronal growth supplement and Pen/Strep solution.
Catalog No. NC0268695 Supplier Sciencell Research Laboratories Supplier No. 1521
Neuronal medium is a complete medium designed for optimal growth of normal human neurons in vitro. It is shipped as a kit with neuronal growth supplement and Pen/Strep solution.