Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates BIOTIN 1/EA
Supplier:  Biotium 290951
Encompass_Preferred
This product is wheat germ agglutinin (WGA) labeled with Biotin WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine WGA conjugates can be used to stain glycoproteins that have these sugar moieties WGA can be used as a gram stain to label gram bacteria but not gram- bacteria WGA also binds to the bud scars on budding yeast such as emSaccharomyces cerevisiae
Catalog No. 50-005-00252