Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates BIOTIN 1/EA
Supplier:  Biotium 29120
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage Sambucus nigra lectin is available conjugated to biotin and a selection of bright and photostable CF Dyes
Catalog No. 50-004-99711