Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates BIOTIN 1/EA
Catalog No. 5000500083
Encompass_Preferred
This product is ulex europaeus agglutinin I (UEA I) labeled with biotin UEA I binds to many glycoproteins and glycolipids containing -linked fucose residues such as ABO blood group glycoconjugates
Catalog No. 50-005-00083 Supplier Biotium Supplier No. 29108
This product is ulex europaeus agglutinin I (UEA I) labeled with biotin UEA I binds to many glycoproteins and glycolipids containing -linked fucose residues such as ABO blood group glycoconjugates