Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates BIOTIN 5/PK
Catalog No. 5000499717
Encompass_Preferred
This product is wheat germ agglutinin (WGA) labeled with Biotin WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine WGA conjugates can be used to stain glycoproteins that have these sugar moieties WGA can be used as a gram stain to label gram bacteria but not gram- bacteria WGA also binds to the bud scars on budding yeast such as emSaccharomyces cerevisiae/em
Catalog No. 50-004-99717 Supplier Biotium Supplier No. 29095
This product is wheat germ agglutinin (WGA) labeled with Biotin WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine WGA conjugates can be used to stain glycoproteins that have these sugar moieties WGA can be used as a gram stain to label gram bacteria but not gram- bacteria WGA also binds to the bud scars on budding yeast such as emSaccharomyces cerevisiae/em