Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF488 1/EA
Catalog No. 5000501232
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Green fluorescent CF488A dye has excitation/emission at 490/515 nm
Catalog No. 50-005-01232 Supplier Biotium Supplier No. 29121
Add to Cart
Add to Cart
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Green fluorescent CF488A dye has excitation/emission at 490/515 nm