Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF532 5/PK
Catalog No. 5000500177
Encompass_Preferred
Wheat germ agglutinin (WGA) labeled with orange fluorescent CF532 dye CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and specificity compared to other dyes CF532 dye has excitation/emission at 527/558 nm 5x1mg
Catalog No. 50-005-00177 Supplier Biotium Supplier No. 29064
Add to Cart
Add to Cart
Wheat germ agglutinin (WGA) labeled with orange fluorescent CF532 dye CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and specificity compared to other dyes CF532 dye has excitation/emission at 527/558 nm 5x1mg