Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF568 1/EA
Catalog No. 5000501245
Encompass_Preferred
This product is fluorescently-labeled datura stramonium lectin (DSL) DSL is a carbohydrate-binding lectin to N-acetylglucosamine and oligomers containing repeating N-acetylglucosamine sequences
Catalog No. 50-005-01245 Supplier Biotium Supplier No. 29098
This product is fluorescently-labeled datura stramonium lectin (DSL) DSL is a carbohydrate-binding lectin to N-acetylglucosamine and oligomers containing repeating N-acetylglucosamine sequences