Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF568 1/EA
Catalog No. 5000501247
Encompass_Preferred
This product is fluorescently-labeled lycopersicon esculentum lectin (tomato) LEL is a marker of blood vessels and microglial cells in rodents
Catalog No. 50-005-01247 Supplier Biotium Supplier No. 29104
Add to Cart
Add to Cart
This product is fluorescently-labeled lycopersicon esculentum lectin (tomato) LEL is a marker of blood vessels and microglial cells in rodents