Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF568 1/EA
Catalog No. 5000500132
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Red fluorescent CF568 dye has excitation/emission at 562/583 nm
Catalog No. 50-005-00132 Supplier Biotium Supplier No. 29122
Add to Cart
Add to Cart
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Red fluorescent CF568 dye has excitation/emission at 562/583 nm