Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF594 1/EA
Catalog No. 5000501222
Encompass_Preferred
This product is fluorescently-labeled ulex europaeus agglutinin I (UEA I) UEA I binds to many glycoproteins and glycolipids containing -linked fucose residues such as ABO blood group glycoconjugates
Catalog No. 50-005-01222 Supplier Biotium Supplier No. 29111
This product is fluorescently-labeled ulex europaeus agglutinin I (UEA I) UEA I binds to many glycoproteins and glycolipids containing -linked fucose residues such as ABO blood group glycoconjugates