Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF594 1/EA
Catalog No. 5000499703
Encompass_Preferred
PHA-L triggers lymphocyte agglutination and is available conjugated to biotin and a selection of 5 bright and photostable CF Dyes Deep-red fluorescent CF594 dye has excitation/emission at 593/614 nm
Catalog No. 50-004-99703 Supplier Biotium Supplier No. 29117
PHA-L triggers lymphocyte agglutination and is available conjugated to biotin and a selection of 5 bright and photostable CF Dyes Deep-red fluorescent CF594 dye has excitation/emission at 593/614 nm