Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF594 1/EA
Catalog No. 5000501310
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Deep-red fluorescent CF594 dye has excitation/emission at 593/614 nm
Catalog No. 50-005-01310 Supplier Biotium Supplier No. 29123
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Deep-red fluorescent CF594 dye has excitation/emission at 593/614 nm