Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF640 1/EA
Catalog No. 5000500239
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Far-red fluorescent CF640R dye has excitation/emission at 642/662 nm
Catalog No. 50-005-00239 Supplier Biotium Supplier No. 29124
Add to Cart
Add to Cart
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Far-red fluorescent CF640R dye has excitation/emission at 642/662 nm