Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF680 1/EA
Supplier:  Biotium 29119
Encompass_Preferred
PHA-L triggers lymphocyte agglutination and is available conjugated to biotin and a selection of 5 bright and photostable CF Dyes
Catalog No. 50-005-00233