Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF680 1/EA
Supplier:  Biotium 29107
Encompass_Preferred
This product is fluorescently-labeled lycopersicon esculentum lectin (tomato) LEL is a marker of blood vessels and microglial cells in rodents
Catalog No. 50-005-01080