Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF680 1/EA
Catalog No. 5000501311
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Far-red/near-IR fluorescent CF680 dye has Ex/Em at 681/698 nm
Catalog No. 50-005-01311 Supplier Biotium Supplier No. 29125
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Far-red/near-IR fluorescent CF680 dye has Ex/Em at 681/698 nm