Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 1/EA
Catalog No. 5000501165
Encompass_Preferred
PHA-L triggers lymphocyte agglutination and is available conjugated to biotin and a selection of 5 bright and photostable CF Dyes Near-infrared CF 740 has Ex/Em at 742/767 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-005-01165 Supplier Biotium Supplier No. 29134
PHA-L triggers lymphocyte agglutination and is available conjugated to biotin and a selection of 5 bright and photostable CF Dyes Near-infrared CF 740 has Ex/Em at 742/767 nm This conjugate is supplied at 1 mg/mL (1 mL size)