Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 1/EA
Catalog No. 5000501167
Encompass_Preferred
This product is fluorescently-labeled datura stramonium lectin (DSL) DSL is a carbohydrate-binding lectin to N-acetylglucosamine and oligomers containing repeating N-acetylglucosamine sequences Near-infrared CF 740 has Ex/Em at 742/767 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-005-01167 Supplier Biotium Supplier No. 29131
This product is fluorescently-labeled datura stramonium lectin (DSL) DSL is a carbohydrate-binding lectin to N-acetylglucosamine and oligomers containing repeating N-acetylglucosamine sequences Near-infrared CF 740 has Ex/Em at 742/767 nm This conjugate is supplied at 1 mg/mL (1 mL size)