Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 1/EA
Supplier:  Biotium 29132
Encompass_Preferred
This product is fluorescently-labeled lycopersicon esculentum lectin (tomato) LEL is a marker of blood vessels and microglial cells in rodents Near-infrared CF 740 has Ex/Em at 742/767 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-005-01168