Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF740 1/EA
Catalog No. 5000501036
Encompass_Preferred
This product is fluorescently-labeled Ulex europaeus agglutinin I (UEA I) UEA I binds to glycoproteins and glycolipids containing -linked fucose residues such as ABO blood group glycoconjugates Near-infrared CF 740 has Ex/Em at 742/767 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-005-01036 Supplier Biotium Supplier No. 29133
Add to Cart
Add to Cart
This product is fluorescently-labeled Ulex europaeus agglutinin I (UEA I) UEA I binds to glycoproteins and glycolipids containing -linked fucose residues such as ABO blood group glycoconjugates Near-infrared CF 740 has Ex/Em at 742/767 nm This conjugate is supplied at 1 mg/mL (1 mL size)