Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 1/EA
Catalog No. 5000501244
Encompass_Preferred
This product is fluorescently-labeled lycopersicon esculentum lectin (tomato) LEL is a marker of blood vessels and microglial cells in rodents Near-infrared CF790 has Ex/Em at 783/808 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-005-01244 Supplier Biotium Supplier No. 29147
This product is fluorescently-labeled lycopersicon esculentum lectin (tomato) LEL is a marker of blood vessels and microglial cells in rodents Near-infrared CF790 has Ex/Em at 783/808 nm This conjugate is supplied at 1 mg/mL (1 mL size)