Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 1/EA
Supplier:  Biotium 29146
Encompass_Preferred
This product is fluorescently-labeled datura stramonium lectin (DSL) DSL is a carbohydrate-binding lectin to N-acetylglucosamine and oligomers containing repeating N-acetylglucosamine sequences Near-infrared CF790 has Ex/Em at 783/808 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-004-99714