Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 1/EA
Catalog No. 5000500069
Encompass_Preferred
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Near-infrared CF790 has Ex/Em at 783/808 nm 1 mg lyophilized solid
Catalog No. 50-005-00069 Supplier Biotium Supplier No. 29150
Sambucus nigra lectin isolated from elderberry bark binds preferentially to sialic acid attached to terminal galactose in -2 6 and to a lesser degree -2 3 linkage It is available conjugated to biotin and CF Dyes Near-infrared CF790 has Ex/Em at 783/808 nm 1 mg lyophilized solid