Learn More
Biotium Neuronal Tracers Fluorescent Proteins Nucleotides Other Conjugates Lectin Conjugates CF790 1/EA
Catalog No. 5000500154
Encompass_Preferred
PHA-L triggers lymphocyte agglutination and is available conjugated to biotin and a selection of 5 bright and photostable CF Dyes Near-infrared CF790 has Ex/Em at 783/808 nm This conjugate is supplied at 1 mg/mL (1 mL size)
Catalog No. 50-005-00154 Supplier Biotium Supplier No. 29149
PHA-L triggers lymphocyte agglutination and is available conjugated to biotin and a selection of 5 bright and photostable CF Dyes Near-infrared CF790 has Ex/Em at 783/808 nm This conjugate is supplied at 1 mg/mL (1 mL size)