Learn More
Apexbio Technology LLC DiscoveryProbe™ Neuronal Signaling Library, 100uL/well(10 mM solution),96 Well Racks With Screw Cap
SDP
Catalog No. 501905591
Encompass_Preferred
A unique collection of 556 neuronal signaling-related small molecules for neuroscience reasearch. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5591 Supplier Apexbio Technology LLC Supplier No. L1026.1
A unique collection of 556 neuronal signaling-related small molecules for neuroscience reasearch. Other sizes are available. Please inqury us for quote.