Learn More
ILEX LIFE SCIENCES LLC NEUROPAN BASAL MEDIUM 500 ML
SDP
Catalog No. 502733904
Encompass_Preferred
PAN-Biotech Neuropan Serum-Free Medium for Neuron Cultivation: Basal Medium, w: L-Glutamine, w/o: Glutamate, w: 2.2 g/L NaHCO3 (500 ml). INTRO: Neuropan Basal Medium is formulated to meet special requirements of neuronal cells. When supplemented with Neuropan-27 Supplement or Neuropan-2 Supplement, it allows long-term maintenance of normal phenotype and growth of neuronal cells, and maintains pure populations of neuronal cells without the need for an astrocyte feeder. APPLICATION: Neuropan Basal Medium meets the special cell culture requirements of prenatal and embryonic neuronal cells from hippocampus, cortex, and other regions of the brain. It allows for both long and short term maintenance of homogenous populations of neuronal cells without the need of an astrocyte feeder layer. STERILE: Yes. STORAGE: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. COMPOSITION: Contains salts, amino acids, glucose, L-Glutamine, and trace elements in an optimized formulation.
Catalog No. 50-273-3904 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0400900
PAN-Biotech Neuropan Serum-Free Medium for Neuron Cultivation: Basal Medium, w: L-Glutamine, w/o: Glutamate, w: 2.2 g/L NaHCO3 (500 ml). INTRO: Neuropan Basal Medium is formulated to meet special requirements of neuronal cells. When supplemented with Neuropan-27 Supplement or Neuropan-2 Supplement, it allows long-term maintenance of normal phenotype and growth of neuronal cells, and maintains pure populations of neuronal cells without the need for an astrocyte feeder. APPLICATION: Neuropan Basal Medium meets the special cell culture requirements of prenatal and embryonic neuronal cells from hippocampus, cortex, and other regions of the brain. It allows for both long and short term maintenance of homogenous populations of neuronal cells without the need of an astrocyte feeder layer. STERILE: Yes. STORAGE: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. COMPOSITION: Contains salts, amino acids, glucose, L-Glutamine, and trace elements in an optimized formulation.